Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGCGCATCCTAGGCGGAGTCAT[A/C]AGCGCCATCAGGTAAGGCGGAGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114170 MIM: 123995 | ||||||||||||||||||||
Literature Links: |
CAPNS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPNS1 - calpain small subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003962.2 | 349 | Silent Mutation | ATA,ATC | I,I 66 | NP_001003962.1 | |
NM_001302632.1 | 349 | Silent Mutation | ATA,ATC | I,I 66 | NP_001289561.1 | |
NM_001302633.1 | 349 | Intron | NP_001289562.1 | |||
NM_001749.3 | 349 | Silent Mutation | ATA,ATC | I,I 66 | NP_001740.1 | |
XM_005259295.1 | 349 | Silent Mutation | ATA,ATC | I,I 66 | XP_005259352.1 | |
XM_005259296.1 | 349 | Silent Mutation | ATA,ATC | I,I 66 | XP_005259353.1 | |
XM_011527359.1 | 349 | Intron | XP_011525661.1 |
COX7A1 - cytochrome c oxidase subunit 7A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |