Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTGACAATTGGTGGAACCATT[G/T]CCCTTTGCATTGGACTTCTTACATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613894 | ||||||||||||||||||||
Literature Links: |
DPY19L3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPY19L3 - dpy-19 like 3 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172774.1 | 339 | Missense Mutation | GCC,TCC | A,S 54 | NP_001166245.1 | |
NM_207325.2 | 339 | Missense Mutation | GCC,TCC | A,S 54 | NP_997208.2 | |
XM_006723045.1 | 339 | Intron | XP_006723108.1 | |||
XM_011526526.2 | 339 | Missense Mutation | GCC,TCC | A,S 54 | XP_011524828.1 | |
XM_017026366.1 | 339 | Missense Mutation | GCC,TCC | A,S 54 | XP_016881855.1 | |
XM_017026367.1 | 339 | UTR 5 | XP_016881856.1 |
LOC400684 - uncharacterized LOC400684 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |