Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGTCACCTCCTTGTACACCAGG[A/C]GGATGATGTGCGGGGGTAGGTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610309 | ||||||||||||||||||||
Literature Links: |
UBE2S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2S - ubiquitin conjugating enzyme E2 S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014501.2 | 598 | Intron | NP_055316.2 | |||
XM_011526752.1 | 598 | Missense Mutation | CGC,CTC | R,L 14 | XP_011525054.1 |