Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCAAGCCGGCCATGCCCATGC[A/G]GATGAGATTCTCTACTGCGTAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRA5 - leukocyte immunoglobulin like receptor A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021250.3 | Intron | NP_067073.1 | ||||
NM_181879.2 | Intron | NP_870994.1 | ||||
NM_181985.3 | Intron | NP_871714.1 | ||||
NM_181986.2 | Intron | NP_871715.1 |