Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGAGCTGGACTTCACGGGGGCC[A/G]GCCGCAAGCACAGCAATGTGAGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606936 | ||||||||||||||||||||
Literature Links: |
TRPM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRPM4 - transient receptor potential cation channel subfamily M member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195227.1 | 358 | Missense Mutation | AGC,GGC | S,G 84 | NP_001182156.1 | |
NM_001321281.1 | 358 | Missense Mutation | AGC,GGC | S,G 84 | NP_001308210.1 | |
NM_001321282.1 | 358 | UTR 5 | NP_001308211.1 | |||
NM_001321283.1 | 358 | Intron | NP_001308212.1 | |||
NM_001321285.1 | 358 | Intron | NP_001308214.1 | |||
NM_017636.3 | 358 | Missense Mutation | AGC,GGC | S,G 84 | NP_060106.2 | |
XM_005259017.1 | 358 | Intron | XP_005259074.1 |