Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACACTGTCATGAATTTTCTTCT[C/T]GGGGCTCTCAGGAACCAATCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611387 MIM: 151750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCL17 - C-X-C motif chemokine ligand 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198477.2 | 396 | Silent Mutation | CCA,CCG | P,P 60 | NP_940879.1 |
LIPE - lipase E, hormone sensitive type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |