Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCGGTGAGTGCGCATGTGCAC[A/G]TTGAGCGAGCTCTTCTGCGTGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC27A5 - solute carrier family 27 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB45 - zinc finger and BTB domain containing 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316978.1 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_001303907.1 | |
NM_001316979.1 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_001303908.1 | |
NM_001316980.1 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_001303909.1 | |
NM_001316981.1 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_001303910.1 | |
NM_001316982.1 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_001303911.1 | |
NM_032792.3 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | NP_116181.1 | |
XM_006723445.3 | 1736 | Silent Mutation | AAC,AAT | N,N 475 | XP_006723508.1 |