Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGACAGGCCTGCCCATGGCCAG[A/G]GACAGCAGGGCCGTGACGATGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD11B1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD11B1L - hydroxysteroid 11-beta dehydrogenase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LONP1 - lon peptidase 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276479.1 | 2439 | Silent Mutation | NP_001263408.1 | |||
NM_001276480.1 | 2439 | Silent Mutation | NP_001263409.1 | |||
NM_004793.3 | 2439 | Silent Mutation | NP_004784.2 | |||
XM_011528441.2 | 2439 | Intron | XP_011526743.1 |
RPL36 - ribosomal protein L36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |