Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAACCAGGACCTTCTCCTGTCA[C/G]TCCAGGGTGTCCCAGACACCTTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606691 MIM: 173850 | ||||||||||||||||||||
Literature Links: |
CEACAM19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM19 - carcinoembryonic antigen related cell adhesion molecule 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127893.1 | 637 | Missense Mutation | CTC,GTC | L,V 53 | NP_001121365.1 | |
NM_020219.3 | 637 | Missense Mutation | CTC,GTC | L,V 53 | NP_064604.2 |
PVR - poliovirus receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |