Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCTCACACGGGAAAAAACCAGG[C/T]TGTTCAGGTCACTAATGGCCTGTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601310 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4A11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP4A11 - cytochrome P450 family 4 subfamily A member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000778.3 | Intron | NP_000769.2 | ||||
NM_001319155.1 | Intron | NP_001306084.1 | ||||
XM_005270539.1 | Intron | XP_005270596.1 | ||||
XM_017000464.1 | Intron | XP_016855953.1 | ||||
XM_017000465.1 | Intron | XP_016855954.1 |
Set Membership: |
HapMap |