Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGGCCCCACTCACCTGGATGAA[C/T]GGGAAGATCAAGCCCACGGTGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC2A5 - solute carrier family 2 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135585.1 | 1514 | Intron | NP_001129057.1 | |||
NM_003039.2 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | NP_003030.1 | |
XM_005263491.3 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_005263548.1 | |
XM_017002133.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857622.1 | |
XM_017002134.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857623.1 | |
XM_017002135.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857624.1 | |
XM_017002136.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857625.1 | |
XM_017002137.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857626.1 | |
XM_017002138.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857627.1 | |
XM_017002139.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857628.1 | |
XM_017002140.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857629.1 | |
XM_017002141.1 | 1514 | Silent Mutation | CCA,CCG | P,P 431 | XP_016857630.1 | |
XM_017002142.1 | 1514 | Silent Mutation | CCA,CCG | P,P 372 | XP_016857631.1 |