Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGGAAGGTGGGGCTGACCGATA[A/G]GACCCTGACCACCTTCATCGAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605708 MIM: 610278 | ||||||||||||||||||||
Literature Links: |
ARHGEF11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF11 - Rho guanine nucleotide exchange factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC71 - leucine rich repeat containing 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144702.2 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | NP_653303.2 | |
XM_005244926.3 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_005244983.1 | |
XM_005244927.3 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_005244984.1 | |
XM_005244928.2 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_005244985.1 | |
XM_006711185.3 | 1046 | Missense Mutation | AAG,AGG | K,R 154 | XP_006711248.1 | |
XM_006711186.3 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_006711249.1 | |
XM_006711187.3 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_006711250.1 | |
XM_011509239.2 | 1046 | Intron | XP_011507541.1 | |||
XM_011509240.2 | 1046 | Intron | XP_011507542.1 | |||
XM_011509241.2 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_011507543.1 | |
XM_017000459.1 | 1046 | Missense Mutation | AAG,AGG | K,R 181 | XP_016855948.1 | |
XM_017000460.1 | 1046 | UTR 5 | XP_016855949.1 | |||
XM_017000461.1 | 1046 | UTR 5 | XP_016855950.1 | |||
XM_017000462.1 | 1046 | UTR 5 | XP_016855951.1 |
PEAR1 - platelet endothelial aggregation receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |