Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 107670 MIM: 147139 MIM: 603881 | ||||||||||||||||||||
Literature Links: |
APOA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOA2 - apolipoprotein A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FCER1G - Fc fragment of IgE receptor Ig | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5187 - microRNA 5187 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR1I3 - nuclear receptor subfamily 1 group I member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM40L - translocase of outer mitochondrial membrane 40 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286373.1 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | NP_001273302.1 | |
NM_001286374.1 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | NP_001273303.1 | |
NM_032174.5 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | NP_115550.2 | |
XM_006711572.2 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | XP_006711635.1 | |
XM_011510057.2 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | XP_011508359.1 | |
XM_017002481.1 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | XP_016857970.1 | |
XM_017002482.1 | 465 | Silent Mutation | CAA,CAG | Q,Q 44 | XP_016857971.1 |