Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATATGTCATGTCCCCTATCTG[A/G]GAAATCTTAAGCAAGAGAAGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEMA4A - semaphorin 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193300.1 | Intron | NP_001180229.1 | ||||
NM_001193301.1 | Intron | NP_001180230.1 | ||||
NM_001193302.1 | Intron | NP_001180231.1 | ||||
NM_022367.3 | Intron | NP_071762.2 | ||||
XM_011509871.2 | Intron | XP_011508173.1 | ||||
XM_011509872.2 | Intron | XP_011508174.1 | ||||
XM_011509873.2 | Intron | XP_011508175.1 | ||||
XM_011509874.2 | Intron | XP_011508176.1 | ||||
XM_011509875.2 | Intron | XP_011508177.1 | ||||
XM_011509876.2 | Intron | XP_011508178.1 | ||||
XM_011509878.2 | Intron | XP_011508180.1 | ||||
XM_011509879.2 | Intron | XP_011508181.1 | ||||
XM_017002056.1 | Intron | XP_016857545.1 | ||||
XM_017002057.1 | Intron | XP_016857546.1 |