Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGAGCAATGCGAGACGGAAAAG[A/G]TTTTTTGCAAGGCTTCCTGTATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608375 MIM: 601007 MIM: 613461 | ||||||||||||||||||||
Literature Links: |
DNAJC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC6 - DnaJ heat shock protein family (Hsp40) member C6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LEPR - leptin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003679.3 | 131 | Intron | NP_001003679.1 | |||
NM_001003680.3 | 131 | Intron | NP_001003680.1 | |||
NM_001198687.1 | 131 | Intron | NP_001185616.1 | |||
NM_001198688.1 | 131 | Intron | NP_001185617.1 | |||
NM_001198689.1 | 131 | Intron | NP_001185618.1 | |||
NM_002303.5 | 131 | Intron | NP_002294.2 |
LEPROT - leptin receptor overlapping transcript | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198681.1 | 131 | Missense Mutation | GAT,GGT | D,G 6 | NP_001185610.1 | |
NM_001198683.1 | 131 | Intron | NP_001185612.1 | |||
NM_017526.4 | 131 | Intron | NP_059996.1 |