Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTAAATAATCACCACACCAAAGT[C/T]CCTCATGTCAAACTGCTTTATTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 139380 MIM: 611616 | ||||||||||||||||||||
Literature Links: |
GNB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNB1 - G protein subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282538.1 | 3046 | UTR 3 | NP_001269467.1 | |||
NM_001282539.1 | 3046 | UTR 3 | NP_001269468.1 | |||
NM_002074.4 | 3046 | UTR 3 | NP_002065.1 | |||
XM_017001059.1 | 3046 | Intron | XP_016856548.1 | |||
XM_017001060.1 | 3046 | Intron | XP_016856549.1 | |||
XM_017001061.1 | 3046 | Intron | XP_016856550.1 |
NADK - NAD kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |