Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTTTTAAGTTTGGCCGCACTG[A/G]AGACTTATGGCAGTGAACACATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 111700 MIM: 605348 | ||||||||||||||||||||
Literature Links: |
RHCE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHCE - Rh blood group CcEe antigens | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM50A - transmembrane protein 50A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014313.3 | 630 | Missense Mutation | GAA,GGA | E,G 153 | NP_055128.1 | |
XM_005245817.1 | 630 | Missense Mutation | GAA,GGA | E,G 122 | XP_005245874.1 | |
XM_011541159.1 | 630 | Missense Mutation | GAA,GGA | E,G 153 | XP_011539461.1 |