Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCTGATTTAGTTATCTCTGATT[A/T]AGTTCCAGCCAAAGAGGAAGCTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600114 | ||||||||||||||||||||
Literature Links: |
CCT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT3 - chaperonin containing TCP1 subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSACC - TSSK6 activating cochaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304817.1 | Intron | NP_001291746.1 | ||||
NM_001304818.1 | Intron | NP_001291747.1 | ||||
NM_001304819.1 | Intron | NP_001291748.1 | ||||
NM_001304820.1 | Intron | NP_001291749.1 | ||||
NM_001304821.1 | Intron | NP_001291750.1 | ||||
NM_001304822.1 | Intron | NP_001291751.1 | ||||
NM_001304823.1 | Intron | NP_001291752.1 | ||||
NM_001304824.1 | Intron | NP_001291753.1 | ||||
NM_001304825.1 | Intron | NP_001291754.1 | ||||
NM_001304826.1 | Intron | NP_001291755.1 | ||||
NM_144627.4 | Intron | NP_653228.1 |