Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAGGACACCCACAACATGTATT[C/T]ATTGATTAAGAACACCCACGACGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCAN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCAN3 - RCAN family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251977.1 | Intron | NP_001238906.1 | ||||
NM_001251978.1 | Intron | NP_001238907.1 | ||||
NM_001251979.1 | Intron | NP_001238908.1 | ||||
NM_001251980.1 | Intron | NP_001238909.1 | ||||
NM_001251981.1 | Intron | NP_001238910.1 | ||||
NM_001251982.1 | Intron | NP_001238911.1 | ||||
NM_001251983.1 | Intron | NP_001238912.1 | ||||
NM_001251984.1 | Intron | NP_001238913.1 | ||||
NM_001251985.1 | Intron | NP_001238914.1 | ||||
NM_013441.3 | Intron | NP_038469.1 |
RCAN3AS - RCAN3 antisense | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |