Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAGGGTACGGACCCCACATGAG[G/T]GTGAGGCTCTCTGCACACTCCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613147 | ||||||||||||||||||||
Literature Links: |
MIR205 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR205 - microRNA 205 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR205HG - MIR205 host gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104548.1 | 546 | Missense Mutation | GAT,TAT | D,Y 55 | NP_001098018.1 | |
XM_017002058.1 | 546 | Missense Mutation | GAT,TAT | D,Y 55 | XP_016857547.1 |