Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACTTCTGATATTTGGCAATCTT[C/T]TCATCCAGTGCTTTAATAACGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602135 MIM: 609365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNALI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNALI1 - dynein axonemal light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNL2 - G protein nucleolar 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323623.1 | 1969 | Silent Mutation | GAA,GAG | E,E 670 | NP_001310552.1 | |
NM_001323624.1 | 1969 | Silent Mutation | GAA,GAG | E,E 419 | NP_001310553.1 | |
NM_013285.2 | 1969 | Silent Mutation | GAA,GAG | E,E 602 | NP_037417.1 |