Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAATTGGAGAGGGAGCCGATGAC[C/T]CGGAAGCGAGAGGGGCTGTGGGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600423 | ||||||||||||||||||||
Literature Links: |
ECE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECE1 - endothelin converting enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113347.1 | 2434 | Silent Mutation | CGA,CGG | R,R 726 | NP_001106818.1 | |
NM_001113348.1 | 2434 | Silent Mutation | CGA,CGG | R,R 722 | NP_001106819.1 | |
NM_001113349.1 | 2434 | Silent Mutation | CGA,CGG | R,R 735 | NP_001106820.1 | |
NM_001397.2 | 2434 | Silent Mutation | CGA,CGG | R,R 738 | NP_001388.1 | |
XM_006710398.2 | 2434 | Silent Mutation | CGA,CGG | R,R 721 | XP_006710461.1 | |
XM_011540872.2 | 2434 | Silent Mutation | CGA,CGG | R,R 746 | XP_011539174.1 | |
XM_011540873.2 | 2434 | Silent Mutation | CGA,CGG | R,R 721 | XP_011539175.1 | |
XM_017000511.1 | 2434 | Silent Mutation | CGA,CGG | R,R 721 | XP_016856000.1 |