Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGATCAGTTTCATCAGAAAAG[A/G]GAAAACTGGTGAGTGTCTAGATATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611673 | ||||||||||||||||||||
Literature Links: |
SWT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105518.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | NP_001098988.1 | |
NM_017673.6 | 322 | Missense Mutation | AGA,GGA | R,G 53 | NP_060143.4 | |
XM_005245271.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_005245328.1 | |
XM_005245273.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_005245330.1 | |
XM_011509657.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_011507959.1 | |
XM_017001527.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_016857016.1 | |
XM_017001528.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_016857017.1 | |
XM_017001529.1 | 322 | UTR 5 | XP_016857018.1 | |||
XM_017001530.1 | 322 | Missense Mutation | AGA,GGA | R,G 53 | XP_016857019.1 |
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |