Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCCAAGGCCCAGGAAGCCAC[A/G]CGGCTGACCCAGCTACCCCACTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEXM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LEXM - lymphocyte expansion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110533.1 | 351 | Silent Mutation | ACA,ACG | T,T 99 | NP_001104003.1 | |
NM_152607.2 | 351 | Silent Mutation | ACA,ACG | T,T 99 | NP_689820.2 | |
XM_017000480.1 | 351 | Silent Mutation | ACA,ACG | T,T 99 | XP_016855969.1 | |
XM_017000481.1 | 351 | Silent Mutation | ACA,ACG | T,T 99 | XP_016855970.1 |
TTC22 - tetratricopeptide repeat domain 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |