Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCATTTTCCTTTGGTTCAGGTG[C/T]GCTGTTGTCCTCCAACTGCAATAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC17 - coiled-coil domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPBP1L1 - GC-rich promoter binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021639.4 | 2323 | Missense Mutation | ACA,GCA | T,A 354 | NP_067652.1 | |
XM_017002019.1 | 2323 | Missense Mutation | ACA,GCA | T,A 301 | XP_016857508.1 |