Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAATGGTGTTCCCTACAGTTGCA[A/G]TCCCAGCATTGCTCCTTGGTGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609209 | ||||||||||||||||||||
Literature Links: |
IVNS1ABP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IVNS1ABP - influenza virus NS1A binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006469.4 | 2290 | Missense Mutation | ACT,ATT | T,I 601 | NP_006460.2 | |
XM_005244843.3 | 2290 | Missense Mutation | ACT,ATT | T,I 383 | XP_005244900.1 | |
XM_011509079.2 | 2290 | Missense Mutation | ACT,ATT | T,I 608 | XP_011507381.1 | |
XM_011509080.2 | 2290 | Missense Mutation | ACT,ATT | T,I 519 | XP_011507382.1 | |
XM_017000084.1 | 2290 | Missense Mutation | ACT,ATT | T,I 390 | XP_016855573.1 |
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |