Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGAGCTGTTGCATGAAGCTGA[C/T]GCCCTCCTCTTCTGGCTCAGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609121 MIM: 603272 MIM: 616698 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPER4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPER4 - cation channel sperm associated 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297647.1 | 427 | Silent Mutation | GAC,GAT | D,D 123 | NP_001284576.1 | |
NM_001297648.1 | 427 | UTR 5 | NP_001284577.1 | |||
NM_006314.2 | 427 | Silent Mutation | GAC,GAT | D,D 123 | NP_006305.2 | |
XM_011540484.2 | 427 | UTR 5 | XP_011538786.1 | |||
XM_011540485.2 | 427 | Intron | XP_011538787.1 |
ZNF593 - zinc finger protein 593 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |