Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACATCACTCACTTGGATCTGGT[C/T]CACAGGCGCCGTCGCTAAATTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606820 MIM: 600063 | ||||||||||||||||||||
Literature Links: |
GLRX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRX2 - glutaredoxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243399.1 | 493 | Missense Mutation | AAC,GAC | N,D 18 | NP_001230328.1 | |
NM_001319291.1 | 493 | Missense Mutation | AAC,GAC | N,D 18 | NP_001306220.1 | |
NM_016066.4 | 493 | Missense Mutation | AAC,GAC | N,D 59 | NP_057150.2 | |
NM_197962.2 | 493 | Missense Mutation | AAC,GAC | N,D 58 | NP_932066.1 | |
XM_017001397.1 | 493 | Missense Mutation | AAC,GAC | N,D 18 | XP_016856886.1 |
TROVE2 - TROVE domain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |