Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAGGATAAATGTTTACGCTTCAC[A/G]TGGCCACCCAGATCCATTTTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608069 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERRFI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERRFI1 - ERBB receptor feedback inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018948.3 | 1613 | Silent Mutation | CAC,CAT | H,H 450 | NP_061821.1 | |
XM_005263477.2 | 1613 | Silent Mutation | CAC,CAT | H,H 399 | XP_005263534.1 | |
XM_006710697.2 | 1613 | Silent Mutation | CAC,CAT | H,H 450 | XP_006710760.1 | |
XM_011541596.2 | 1613 | Silent Mutation | CAC,CAT | H,H 375 | XP_011539898.1 |