Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGCCTGCGGCCCGGGGGCCCAG[C/G]AGGGGACGGCAGAGCAGAGGTCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616824 | ||||||||||||||||||||
Literature Links: |
TRNP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRNP1 - TMF1-regulated nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013642.2 | 126 | Missense Mutation | CAG,GAG | Q,E 15 | NP_001013664.1 | |
XM_005245867.3 | 126 | Missense Mutation | CAG,GAG | Q,E 15 | XP_005245924.1 |