Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCCGGGGCGGTGATTGCAGCC[C/T]CAGACAGCCGGCGCTGGCTGTGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604376 | ||||||||||||||||||||
Literature Links: |
MPZL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPZL1 - myelin protein zero like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146191.1 | 242 | Missense Mutation | CCA,TCA | P,S 14 | NP_001139663.1 | |
NM_003953.5 | 242 | Missense Mutation | CCA,TCA | P,S 14 | NP_003944.1 | |
NM_024569.4 | 242 | Missense Mutation | CCA,TCA | P,S 14 | NP_078845.3 |