Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTAGGACTGTACCGTACAGGGA[A/C]ATCCTACTTGATGAACCATCTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612467 | ||||||||||||||||||||
Literature Links: |
GBP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBP6 - guanylate binding protein family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320257.1 | 255 | Intron | NP_001307186.1 | |||
NM_198460.2 | 255 | Missense Mutation | AAA,ACA | K,T 51 | NP_940862.2 | |
XM_011540835.2 | 255 | Missense Mutation | AAA,ACA | K,T 51 | XP_011539137.1 |