Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACACACATATTTAATTACATGG[T/G]TAAAGACTATTATTTCGCACACTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607399 | ||||||||||||||||||||
Literature Links: |
MCOLN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCOLN2 - mucolipin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153259.3 | 2113 | UTR 3 | NP_694991.2 | |||
XM_005270719.3 | 2113 | UTR 3 | XP_005270776.1 | |||
XM_006710552.2 | 2113 | UTR 3 | XP_006710615.1 | |||
XM_011541187.2 | 2113 | UTR 3 | XP_011539489.1 | |||
XM_011541188.2 | 2113 | UTR 3 | XP_011539490.1 | |||
XM_017000923.1 | 2113 | Intron | XP_016856412.1 |