Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGATCTCCATCCTTGCCGTTGATT[G/T]CCTGGCCAGGCCGGCCAGGGAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120260 MIM: 606480 | ||||||||||||||||||||
Literature Links: |
COL9A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL9A2 - collagen type IX alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001852.3 | 2033 | Missense Mutation | GAA,GCA | E,A 632 | NP_001843.1 | |
XM_006710365.3 | 2033 | Missense Mutation | GAA,GCA | E,A 632 | XP_006710428.1 | |
XM_011540715.2 | 2033 | Missense Mutation | GAA,GCA | E,A 542 | XP_011539017.1 | |
XM_011540716.2 | 2033 | Missense Mutation | GAA,GCA | E,A 542 | XP_011539018.1 | |
XM_011540717.2 | 2033 | Missense Mutation | GAA,GCA | E,A 451 | XP_011539019.1 | |
XM_017000332.1 | 2033 | Missense Mutation | GAA,GCA | E,A 636 | XP_016855821.1 | |
XM_017000333.1 | 2033 | Missense Mutation | GAA,GCA | E,A 538 | XP_016855822.1 |
ZMPSTE24 - zinc metallopeptidase STE24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |