Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCACATCCCCACAGGCCCTGG[A/C]AGAGTGTCTTGCACACAGTGGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120326 MIM: 610033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL16A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL16A1 - collagen type XVI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001856.3 | Intron | NP_001847.3 | ||||
XM_005270481.1 | Intron | XP_005270538.1 | ||||
XM_011540722.1 | Intron | XP_011539024.1 | ||||
XM_011540723.1 | Intron | XP_011539025.1 | ||||
XM_011540724.1 | Intron | XP_011539026.1 | ||||
XM_011540726.2 | Intron | XP_011539028.1 | ||||
XM_011540727.2 | Intron | XP_011539029.1 | ||||
XM_011540728.2 | Intron | XP_011539030.1 | ||||
XM_011540729.1 | Intron | XP_011539031.1 | ||||
XM_011540730.1 | Intron | XP_011539032.1 | ||||
XM_017000338.1 | Intron | XP_016855827.1 | ||||
XM_017000339.1 | Intron | XP_016855828.1 | ||||
XM_017000340.1 | Intron | XP_016855829.1 | ||||
XM_017000341.1 | Intron | XP_016855830.1 |
PEF1 - penta-EF-hand domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |