Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATCCCAGTGCTAGGATAATGAC[A/C]TTTTATCCAACTATGGAAGAGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609764 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM4A - lysine demethylase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014663.2 | 251 | Silent Mutation | ACA,ACC | T,T 16 | NP_055478.2 | |
XM_005271354.3 | 251 | Silent Mutation | ACA,ACC | T,T 16 | XP_005271411.1 | |
XM_005271355.3 | 251 | Silent Mutation | ACA,ACC | T,T 16 | XP_005271412.1 | |
XM_005271356.3 | 251 | UTR 5 | XP_005271413.1 | |||
XM_011542467.2 | 251 | Intron | XP_011540769.2 | |||
XM_017002912.1 | 251 | UTR 5 | XP_016858401.1 |