Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAATTGGTCCTAAGGCAGCAAAG[A/G]GCCCTGTGGTAAGAAAGAGCATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR14I1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR14I1 - olfactory receptor family 14 subfamily I member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004734.1 | 896 | Missense Mutation | CCC,CTC | P,L 254 | NP_001004734.1 | |
XM_005273132.3 | 896 | Missense Mutation | CCC,CTC | P,L 254 | XP_005273189.1 |