Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTAGAGACTCATTCATCAGCTT[A/G]TTGATCCTTTCTGTATTGGCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FH - fumarate hydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000143.3 | 1841 | Silent Mutation | AAC,AAT | N,N 446 | NP_000134.2 | |
XM_011544132.2 | 1841 | Silent Mutation | AAC,AAT | N,N 370 | XP_011542434.1 |