Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCTAGGAGCGGGGTGCCCTGA[A/G]CTTTGAGCGGCGCTACCATGTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDTC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDTC1 - WD and tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276252.1 | 545 | Missense Mutation | AAC,AGC | N,S 22 | NP_001263181.1 | |
NM_015023.4 | 545 | Missense Mutation | AAC,AGC | N,S 22 | NP_055838.2 | |
XM_011541057.1 | 545 | Missense Mutation | AAC,AGC | N,S 22 | XP_011539359.1 | |
XM_011541058.2 | 545 | Missense Mutation | AAC,AGC | N,S 22 | XP_011539360.1 | |
XM_017000730.1 | 545 | Missense Mutation | AAC,AGC | N,S 22 | XP_016856219.1 |