Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGCCAGTCGGGCTTCATCCAG[T/C]TCCCGCTCAGTAGAGGCCAGCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605313 | ||||||||||||||||||||
Literature Links: |
LIX1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIX1L - limb and CNS expressed 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153713.2 | 995 | Silent Mutation | GAA,GAG | E,E 301 | NP_714924.1 | |
XM_017000316.1 | 995 | Silent Mutation | GAA,GAG | E,E 269 | XP_016855805.1 |
RBM8A - RNA binding motif protein 8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |