Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTCCTGGAAATATTCCTCCTC[C/T]TCATCGAAGAAGCCGTTCTCCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM89A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM89A - family with sequence similarity 89 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198552.2 | 470 | Silent Mutation | GAA,GAG | E,E 144 | NP_940954.1 |
MIR1182 - microRNA 1182 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |