Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTACCTGGAGGGACCTCCCGAG[A/G]GGACTCGAGTGAGCAGGGCAGGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609294 | ||||||||||||||||||||
Literature Links: |
GABPB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABPB2 - GA binding protein transcription factor beta subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA6C - semaphorin 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178061.1 | 3915 | Missense Mutation | CCC,CTC | P,L 900 | NP_001171532.1 | |
NM_001178062.1 | 3915 | Missense Mutation | CCC,CTC | P,L 860 | NP_001171533.1 | |
NM_030913.4 | 3915 | Missense Mutation | CCC,CTC | P,L 868 | NP_112175.2 | |
XM_005244835.3 | 3915 | Missense Mutation | CCC,CTC | P,L 900 | XP_005244892.1 | |
XM_017000075.1 | 3915 | Missense Mutation | CCC,CTC | P,L 900 | XP_016855564.1 | |
XM_017000076.1 | 3915 | Missense Mutation | CCC,CTC | P,L 900 | XP_016855565.1 | |
XM_017000077.1 | 3915 | Missense Mutation | CCC,CTC | P,L 900 | XP_016855566.1 | |
XM_017000078.1 | 3915 | Missense Mutation | CCC,CTC | P,L 868 | XP_016855567.1 | |
XM_017000079.1 | 3915 | Missense Mutation | CCC,CTC | P,L 868 | XP_016855568.1 | |
XM_017000080.1 | 3915 | Missense Mutation | CCC,CTC | P,L 860 | XP_016855569.1 | |
XM_017000081.1 | 3915 | Missense Mutation | CCC,CTC | P,L 860 | XP_016855570.1 | |
XM_017000082.1 | 3915 | Missense Mutation | CCC,CTC | P,L 635 | XP_016855571.1 |