Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCAGCCTGCCAGTCACAAGAA[C/T]GAGCGCTCCCAGCCAGAGGTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GREM2 - gremlin 2, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022469.3 | 1272 | UTR 3 | NP_071914.3 | |||
XM_005273226.3 | 1272 | UTR 3 | XP_005273283.1 | |||
XM_011544249.2 | 1272 | UTR 3 | XP_011542551.1 |