Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCACCAATATGAAACCAACTATC[A/G]TACTTAAAACCTAGGAAATATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD45 - ankyrin repeat domain 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198493.2 | 1549 | UTR 3 | NP_940895.1 | |||
XM_011509466.2 | 1549 | Intron | XP_011507768.1 | |||
XM_011509467.1 | 1549 | Intron | XP_011507769.1 | |||
XM_011509468.1 | 1549 | Intron | XP_011507770.1 | |||
XM_011509470.1 | 1549 | Intron | XP_011507772.1 | |||
XM_011509471.2 | 1549 | Intron | XP_011507773.1 | |||
XM_017001123.1 | 1549 | Intron | XP_016856612.1 | |||
XM_017001124.1 | 1549 | Intron | XP_016856613.1 |
LOC730159 - uncharacterized LOC730159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195190.1 | 1549 | Intron | NP_001182119.1 | |||
XM_011509968.2 | 1549 | Intron | XP_011508270.1 |
SLC9C2 - solute carrier family 9 member C2 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |