Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATGGACTGAGCACAGATATTT[A/G]GGGCTTTCTTGTGTTAGAAAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VASH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VASH2 - vasohibin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136474.2 | Intron | NP_001129946.1 | ||||
NM_001136475.2 | Intron | NP_001129947.1 | ||||
NM_001301056.1 | Intron | NP_001287985.1 | ||||
NM_024749.4 | Intron | NP_079025.2 | ||||
XM_011509987.2 | Intron | XP_011508289.1 | ||||
XM_011509988.2 | Intron | XP_011508290.1 | ||||
XM_011509989.2 | Intron | XP_011508291.1 | ||||
XM_011509991.2 | Intron | XP_011508293.2 | ||||
XM_011509994.1 | Intron | XP_011508296.1 | ||||
XM_011509995.1 | Intron | XP_011508297.1 | ||||
XM_011509997.1 | Intron | XP_011508299.1 | ||||
XM_017002351.1 | Intron | XP_016857840.1 | ||||
XM_017002352.1 | Intron | XP_016857841.1 |