Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTCTGCTGGACGTGTTCTGTAT[A/G]CGGGTGACGCCGTGCCCGGATAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602024 MIM: 610692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf64 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf64 - chromosome 1 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCNKA - chloride voltage-gated channel Ka | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPB7 - heat shock protein family B (small) member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014424.4 | 1293 | Missense Mutation | CAT,TAT | H,Y 156 | NP_055239.1 | |
XM_011541249.1 | 1293 | Missense Mutation | CAT,TAT | H,Y 249 | XP_011539551.1 | |
XM_011541250.2 | 1293 | Missense Mutation | CAT,TAT | H,Y 248 | XP_011539552.1 |