Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTATGTGGCCTTGTCAGGTAT[C/T]CTGCCAGGTAAAATGGGGATAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1522 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1522 - KIAA1522 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YARS - tyrosyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003680.3 | Intron | NP_003671.1 | ||||
XM_011542347.1 | Intron | XP_011540649.1 | ||||
XM_011542348.1 | Intron | XP_011540650.1 | ||||
XM_017002651.1 | Intron | XP_016858140.1 |