Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGCTGACATATAGTAGATGCT[C/T]ATAACTGTTAAAAAGGGGCCAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615100 MIM: 601968 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTTNBP2NL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTTNBP2NL - CTTNBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4256 - microRNA 4256 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WNT2B - Wnt family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291880.1 | 153 | UTR 5 | NP_001278809.1 | |||
NM_004185.4 | 153 | UTR 5 | NP_004176.2 | |||
NM_024494.2 | 153 | Intron | NP_078613.1 |