Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTATTCTCTTCTTCTTGGGGAT[A/G]TCAATTCCTGTTCTGAGCCCGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAPLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAPLN2 - hyaluronan and proteoglycan link protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021817.2 | 335 | Intron | NP_068589.1 | |||
XM_005245415.2 | 335 | Intron | XP_005245472.1 | |||
XM_011509853.2 | 335 | UTR 5 | XP_011508155.1 | |||
XM_017002020.1 | 335 | Intron | XP_016857509.1 | |||
XM_017002021.1 | 335 | Intron | XP_016857510.1 |
LOC101928177 - uncharacterized LOC101928177 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |