Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGAAATTTTGAAGAGAGTCTAA[A/G]GAATGTAGGAAATTAAGGTCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603275 MIM: 600607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP5K1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIP5K1A - phosphatidylinositol-4-phosphate 5-kinase type 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135636.1 | Intron | NP_001129108.1 | ||||
NM_001135637.1 | Intron | NP_001129109.1 | ||||
NM_001135638.1 | Intron | NP_001129110.1 | ||||
NM_003557.2 | Intron | NP_003548.1 | ||||
XM_005245525.4 | Intron | XP_005245582.1 | ||||
XM_005245527.3 | Intron | XP_005245584.1 | ||||
XM_005245528.4 | Intron | XP_005245585.1 | ||||
XM_005245530.4 | Intron | XP_005245587.1 | ||||
XM_005245531.3 | Intron | XP_005245588.1 | ||||
XM_005245532.3 | Intron | XP_005245589.1 | ||||
XM_006711563.3 | Intron | XP_006711626.1 | ||||
XM_006711564.3 | Intron | XP_006711627.1 | ||||
XM_006711567.3 | Intron | XP_006711630.1 | ||||
XM_006711568.3 | Intron | XP_006711631.2 | ||||
XM_011510043.2 | Intron | XP_011508345.1 | ||||
XM_011510045.2 | Intron | XP_011508347.1 | ||||
XM_011510046.2 | Intron | XP_011508348.1 | ||||
XM_011510048.2 | Intron | XP_011508350.1 | ||||
XM_011510049.2 | Intron | XP_011508351.1 | ||||
XM_017002439.1 | Intron | XP_016857928.1 | ||||
XM_017002440.1 | Intron | XP_016857929.1 | ||||
XM_017002441.1 | Intron | XP_016857930.1 |
VPS72 - vacuolar protein sorting 72 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |